EMAIL THIS PAGE TO A FRIEND
Microcirculation (New York, N.Y. : 1994)
Targeting Adenosine Monophosphate-Activated Protein Kinase by Metformin Adjusts Post-Ischemic Hyperemia and Extracellular Neuronal Discharge in Transient Global Cerebral Ischemia.
PMID 26213885