EMAIL THIS PAGE TO A FRIEND
Phytopathology
In Vitro Transcripts of Wild-Type and Fluorescent Protein-Tagged Triticum mosaic virus (Family Potyviridae) are Biologically Active in Wheat.
PMID 26214124
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
791733
Potassium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
C2H3KO2