EMAIL THIS PAGE TO A FRIEND
Nature communications
A biphasic epigenetic switch controls immunoevasion, virulence and niche adaptation in non-typeable Haemophilus influenzae.
PMID 26215614
Abstract
Non-typeable Haemophilus influenzae contains an N(6)-adenine DNA-methyltransferase (ModA) that is subject to phase-variable expression (random ON/OFF switching). Five modA alleles, modA2, modA4, modA5, modA9 and modA10, account for over two-thirds of clinical otitis media isolates surveyed. Here, we use single molecule, real-time (SMRT) methylome analysis to identify the DNA-recognition motifs for all five of these modA alleles. Phase variation of these alleles regulates multiple proteins including vaccine candidates, and key virulence phenotypes such as antibiotic resistance (modA2, modA5, modA10), biofilm formation (modA2) and immunoevasion (modA4). Analyses of a modA2 strain in the chinchilla model of otitis media show a clear selection for ON switching of modA2 in the middle ear. Our results indicate that a biphasic epigenetic switch can control bacterial virulence, immunoevasion and niche adaptation in an animal model system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N8285
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
C21H27N7O14P2
N8410
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
C21H27N7O14P2
N8535
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
C21H27N7O14P2
N7004
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
C21H27N7O14P2 · xH2O
N6522
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast
C21H27N7O14P2 · xH2O
N1636
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
C21H27N7O14P2 · xH2O
N3014
β-Nicotinamide adenine dinucleotide hydrate, cell culture tested, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
C21H27N7O14P2 · xH2O
N7381
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1, ≥95% (HPLC)
C21H27N7O14P2 · xH2O
N1511
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
C21H27N7O14P2 · xH2O
43410
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
C21H27N7O14P2 · xH2O
N7132
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
C21H26LiN7O14P2
51280
Hemin, BioXtra, from Porcine, ≥97.0% (HPLC)
C34H32ClFeN4O4
H9039
Hemin, from bovine, ≥90%
C34H32ClFeN4O4
O1376
Orotidine 5′-monophosphate trisodium salt, ≥99% (HPLC), powder
C10H10N2Na3O11P