PloS one
Reduced Let-7a Is Associated with Chemoresistance in Primary Breast Cancer.
PMID 26218285
Abstract
Chemotherapy resistance remains an important problem in the breast cancer clinic. The ability to predict the patients who would respond to a distinct therapy would help to optimize tailored treatment options. miRNAs can mediate a number of genes in response to drug-induced acute cellular stress. Several studies suggest that let-7 miRNA may be involved in the chemosensitivity of cancer cell lines in vitro. However, it is not known whether this phenomenon occurs in clinical breast tumors. The present study showed that lower let-7a expression was associated with epirubicin resistance in primary breast tumors. Moreover, upregulation of let-7a expression sensitized resistant breast tumor cell lines to epirubicin by enhancing cellular apoptosis in vitro. Collectively, these findings indicate that lower expression of let-7a miRNA can induce chemoresistance in breast cancer by enhancing cellular apoptosis and suggest that let-7a may be used as a therapeutic target to modulate epirubicin-based chemotherapy resistance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D6770
Digoxin, analytical standard
C41H64O14