PloS one
The Rapid Screening of Triazophos Residues in Agricultural Products by Chemiluminescent Enzyme Immunoassay.
PMID 26218576
Abstract
A highly sensitive chemiluminescent enzyme immunoassay (CLEIA) method was developed in this study for efficient screening of triazophos residues in a large number of samples. Based on the maximum residue limits (MRLs) set by China and CAC for triazophos in different agro-products, the representative apple, orange, cabbage, zucchini, and rice samples were selected as spiked samples, and the triazophos at the concentrations of the MRL values were spiked to blank samples. Subsequently, the five samples with the spiked triazophos standard were measured by CLEIA 100 times, and the detection results indicated that the correction factors of the apple, orange, cabbage, zucchini, and rice were determined as 0.79, 0.66, 0.85, 0.76, and 0.91, respectively. In this experiment, 1500 real samples were detected by both the CLEIA and the GC-MS methods. With the GC-MS method, 1462 samples were identified as negative samples and 38 samples as positive samples. Based on the correction factors, the false positive rate of the CLEIA method was 0.13%, and false negative rate was 0. The results showed that the established CLEIA method could be used to screen a large number of real samples.