EMAIL THIS PAGE TO A FRIEND
FEMS microbiology ecology
Elevated CO2 induces a bloom of microphytobenthos within a shell gravel mesocosm.
PMID 26220309
Abstract
The geological storage of carbon dioxide (CO2) is expected to be an important component of future global carbon emission mitigation, but there is a need to understand the impacts of a CO2 leak on the marine environment and to develop monitoring protocols for leakage detection. In the present study, sediment cores were exposed to CO2-acidified seawater at one of five pH levels (8.0, 7.5, 7.0, 6.5 and 6.0) for 10 weeks. A bloom of Spirulina sp. and diatoms appeared on sediment surface exposed to pH 7.0 and 7.5 seawater. Quantitative PCR measurements of the abundance of 16S rRNA also indicated an increase within the pH 7.0 and 7.5 treatments after 10 weeks incubation. More detailed analysis of the microbial communities from the pH 7.0, 7.5 and 8.0 treatments confirmed an increase in the relative abundance of Spirulina sp. and Navicula sp. sequences, with changes in the relative abundance of major archaeal and bacterial groups also detected within the pH 7.0 treatment. A decreased flux of silicate from the sediment at this pH was also detected. Monitoring blooms of microphytobenthos may prove useful as an indicator of CO2 leakage within coastal areas.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
413445
Mercury(II) chloride, ≥98%
Cl2Hg
B81255
N-Bromosuccinimide, ReagentPlus®, 99%
C4H4BrNO2