EMAIL THIS PAGE TO A FRIEND
International journal of clinical and experimental medicine
The 5-hydroxytryptamine transporter is functional in human coronary artery smooth muscle cells proliferation and is regulated by Interleukin-1 beta.
PMID 26221231
Abstract
Abnormal human coronary artery smooth muscle cells (hCASMCs) proliferation and migration are key factors in coronary artery restenosis after percutaneous coronary intervention. Platelets release 5-hydroxytryptamine (5-HT), which is a strong mitogen for pulmonary artery smooth muscle cells proliferation and migration. Here, we investigated the effects of 5-HT and role of 5-HT transporter (5-HTT) on hCASMCs proliferation and migration. The 5-HT (10(-6)-10(-5) mol/l) significantly increased hCASMCs proliferation and migration, and these effects were inhibited by fluoxetine (10(-5) mol/l) and citalopram (10(-6) mol/l), two 5-HTT blocker. Overexpression in hCASMCs enhanced 5-HT induced cells proliferation and migration. The 5-HTT and interleukin-1 beta (IL-1β) expression were increased in rat balloon injury carotid arteries. Treatment with IL-1β (10 ng/ml, 3d) upregulates 5-HTT expression in hCASMCs and increased 5-HT induced currents in Human Embryonic Kidney 293-5-HTT cells.