EMAIL THIS PAGE TO A FRIEND
BioMed research international
Antioxidative Role of Hatikana (Leea macrophylla Roxb.) Partially Improves the Hepatic Damage Induced by CCl4 in Wistar Albino Rats.
PMID 26221590
Abstract
This research investigated the protective role of Leea macrophylla extract on CCl4-induced acute liver injury in rats. Different fractions of Leea macrophylla (Roxb.) crude extract were subjected to analysis for antioxidative effects. Rats were randomly divided into four groups as normal control, hepatic control, and reference control (silymarin) group and treatment group. Evaluations were made for the effects of the fractions on serum enzymes and biochemical parameters of CCl4-induced albino rat. Histopathological screening was also performed to evaluate the changes of liver tissue before and after treatment. Different fractions of Leea macrophylla showed very potent 2,2-diphenyl-1-picrylhydrazyl (DPPH) radical scavenging effect, FeCl3 reducing effect, superoxide scavenging effect, and iron chelating effect. Carbon tetrachloride induction increased the level of serum aspartate aminotransferase (AST), alanine aminotransferase (ALT), and alkaline phosphatase (ALP) and other biochemical parameters such as lipid profiles, total protein, and CK-MB. In contrast, treatment of Leea macrophylla reduced the serum aspartate aminotransferase (AST), alanine aminotransferase (ALT), and alkaline phosphatase (ALP) activities as well as biochemical parameters activities. L. macrophylla partially restored the lipid profiles, total protein, and CK-MB. Histopathology showed the treated liver towards restoration. Results evidenced that L. macrophylla can be prospective source of hepatic management in liver injury.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
281689
1,1-Diphenyl-2-picrylhydrazine, 97%
C18H13N5O6
D9132
2,2-Diphenyl-1-picrylhydrazyl
C18H12N5O6
C7727
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
C21H20O6
C1386
Curcumin, from Curcuma longa (Turmeric), powder
C21H20O6
561932
Deblock trichloroacetic acid solution, 3 % (w/v) in methylene chloride
C2HCl3O2
451649
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Cl3Fe
701122
Iron(III) chloride, sublimed grade, ≥99.9% trace metals basis
Cl3Fe
361003
Iron(III) chloride on silica gel, powder, extent of labeling: 5 wt. % loading
Cl3Fe
710857
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran
Cl3Fe
84010
NBT Vials, for in vitro diagnostic use
C40H30Cl2N10O6
N6876
Nitrotetrazolium Blue chloride, ~98% (TLC)
C40H30Cl2N10O6
N6639
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
C40H30Cl2N10O6
74032
Nitrotetrazolium Blue chloride, ≥98.0% (calc. based on dry substance, T)
C40H30Cl2N10O6
244856
Potassium, ingot, 99.95% trace metals basis
K
244864
Potassium, chunks (in mineral oil), 98% trace metals basis
K
679909
Potassium, cubes (in mineral oil), L × W × H 40 mm × 30 mm × 20 mm, 99.5% trace metals basis
K
215813
Potassium hydride, 30 wt % dispersion in mineral oil
HK
708860
Potassium hydride, in paraffin
HK
190780
Sodium trichloroacetate, 97%
C2Cl3NaO2
672920
TCA Deblock, 3 % (w/v) in methylene chloride
C2HCl3O2
T9159
Trichloroacetic acid, BioXtra, ≥99.0%
C2HCl3O2
T6399
Trichloroacetic acid, ACS reagent, ≥99.0%
C2HCl3O2
T4885
Trichloroacetic acid, ≥99.0% (titration)
C2HCl3O2
T8657
Trichloroacetic acid, for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
C2HCl3O2
91228
Trichloroacetic acid, BioUltra, ≥99.5% (T)
C2HCl3O2
91230
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
C2HCl3O2
27242
Trichloroacetic acid, puriss., meets analytical specification of Ph. Eur., USP 21, 99-100.5% (calc. to the dried substance)
C2HCl3O2
116114
Trichloroacetic acid, reagent grade, 98%
C2HCl3O2
T0324
Trichloroacetic acid solution, ~6.25 % (w/v), 0.38 N
C2HCl3O2
T0699
Trichloroacetic acid solution, 6.1 N
C2HCl3O2