EMAIL THIS PAGE TO A FRIEND
Journal of immunology research
Branched Polyethylenimine-Superparamagnetic Iron Oxide Nanoparticles (bPEI-SPIONs) Improve the Immunogenicity of Tumor Antigens and Enhance Th1 Polarization of Dendritic Cells.
PMID 26221615
Abstract
Nanoparticles in the field of dendritic cell (DC) research are emerging as a promising method of enhancing the efficacy of cancer immunotherapy. We investigated the effect of branched polyethylenimine-superparamagnetic iron oxide nanoparticles (bPEI-SPIONs) on tumor cells loaded onto DCs. The tumor antigens were prepared as follows: (1) apoptotic U266 cells with ultraviolet B (UVB) irradiation followed by a 2 h incubation in the absence (2 h postirradiated cells) or (2) presence of bPEI-SPIONs (bPEI-SPION 2 h postirradiated cells) and (3) apoptotic U266 cells with UVB irradiation followed by an overnight 16 h incubation (16 h postirradiated cells). bPEI-SPIONs render U266 cells sensitive to UVB irradiation through reactive oxygen species production to accelerate apoptotic death. The 2 h postirradiated cells and bPEI-SPION 2 h postirradiated cells released immunogenic proteins, including Hsp70, Hsp90, and HMGB1. The DCs loaded with bPEI-SPION 2 h postirradiated cells showed the highest IL-12p70 production and Th1 polarization compared with other DCs. These results suggest that bPEI-SPIONs are a promising method of enhancing the immunogenicity of tumor cells and promoting Th1 polarization of DCs loaded with these tumor cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B7651
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
C16H24O4
B6542
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
C16H24O4
B5936
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO, 0.2 μm filtered
C16H24O4
SRP4137 IL-4 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
SRP6172
IL-4 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
SRP3093 IL-4 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
I4269 Interleukin-4 human, IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
H7291 Interleukin-4 human, potency:, IL-4, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
I2661 Interleukin-4 human, IL-4, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), animal component free
A7250
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
C5H9NO3S
A5099
N-Acetyl-L-cysteine, PharmaGrade, Ajinomoto, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
C5H9NO3S
A8199
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
C5H9NO3S
A9165
N-Acetyl-L-cysteine, cell culture tested, BioReagent
C5H9NO3S
P6061
Phenylacetic acid, plant cell culture tested
C8H8O2
P16621
Phenylacetic acid, 99%
C8H8O2
W287814
Phenylacetic acid, natural, ≥99%
C8H8O2
W287806
Phenylacetic acid, ≥99%, FCC, FG
C8H8O2
79346
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
C36H56O8
P5592
Potassium peroxodisulfate, BioXtra, ≥99.0%
K2O8S2
379824
Potassium persulfate, 99.99% trace metals basis
K2O8S2
P2713
Potassium thiocyanate, BioXtra, ≥99.0%
CKNS