EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Germline HABP2 Mutation Causing Familial Nonmedullary Thyroid Cancer.
PMID 26222560
Abstract
Familial nonmedullary thyroid cancer accounts for 3 to 9% of all cases of thyroid cancer, but the susceptibility genes are not known. Here, we report a germline variant of HABP2 in seven affected members of a kindred with familial nonmedullary thyroid cancer and in 4.7% of 423 patients with thyroid cancer. This variant was associated with increased HABP2 protein expression in tumor samples from affected family members, as compared with normal adjacent thyroid tissue and samples from sporadic cancers. Functional studies showed that HABP2 has a tumor-suppressive effect, whereas the G534E variant results in loss of function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU051611
MISSION® esiRNA, targeting human HABP2