EMAIL THIS PAGE TO A FRIEND
Journal of aquatic animal health
Influence of Dietary Ascorbic Acid on the Immune Responses of Juvenile Korean Rockfish Sebastes schlegelii.
PMID 26223304
Abstract
Juvenile Korean Rockfish Sebastes schlegelii (length, 13.6 ± 1.4 cm [mean ± SD]; weight, 53.6 ± 4.2 g) were fed twice daily with diets containing varying levels of ascorbic acid (0, 50, 100, 200, and 400 mg/kg) for 4 weeks. Significant increases in daily weight and length occurred in fish fed more than 50 mg/kg ascorbic acid. The lysozyme activity of fish fed diets containing ascorbic acid was considerably increased in rockfish plasma at 400 mg/kg ascorbic acid and in kidney at over 50 mg/kg ascorbic acid. Total plasma immunoglobulin M (IgM) levels were markedly elevated in fish fed 400 mg/kg ascorbic acid. The results suggest that dietary ascorbic acid supplementation in juvenile rockfish can induce a significant increase in growth and in immunological features such as lysozyme activity and plasma IgM levels.