EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
UPLC-ESI-TOF MS-Based Metabolite Profiling of the Antioxidative Food Supplement Garcinia buchananii.
PMID 26226176
Abstract
Comparative antioxidative analyses of aqueous ethanolic extracts from leaf, root, and stem of Garcinia buchananii revealed high activity of all three organs. To investigate the metabolite composition of the different parts of G. buchananii, an untargeted metabolomics approach using UPLC-ESI-TOF MS with simultaneous acquisition of low- and high-collision energy mass spectra (MS(e)) was performed. Unsupervised statistics (PCA) highlighted clear differences in the metabolomes of the three organs. OPLS-DA revealed (2R,3S,2″R,3″R)-GB-1, (2R,3S)-morelloflavone, and (2R,3S)-volkensiflavone as the most decisive marker compounds discriminating leaf from root and stem extract. Leaves represent the best source to isolate GB-1, morelloflavone, and volkensiflavone. Root extract is the best organ to isolate xanthones and stem bark extract the best source to isolate (2R,3S,2″R,3″R)-manniflavanone; the identified polyisoprenylated benzophenones are characteristic compounds for the leaf organ. Morelloflavone, volkensiflavone, and garcicowin C were isolated for the first time from G. buchananii, identified via MS, NMR, and CD spectroscopy, and showed in H2O2 scavenging, H/L-TEAC, and H/L-ORAC assays moderate to strong in vitro antioxidative activities.