EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Iron nanotube-silica composite (ZVI-S-PCAT modified silica composite) preparation, characterization and application as a recyclable catalytic system for 5-membered ring organic transformations.
PMID 26228908
Abstract
Iron nanotube silica composite have been synthesized and studied through particle-size analysis, FTIR, SEM-EDX, TEM, XRD, UV, VSM, TGA-DTA and XPS techniques. The application of nanoframeworks as sustainable recyclable catalytic systems has been observed for azole cyclic ring organic transformations. The good reaction yields and characterization through (1)H NMR, (13)C NMR and mass analysis support the performance of the nanoframeworks. We also present here the synthesis of two novel compounds. Also the prepared nanoframework has been observed to show soft magnetism which provides a scope to be used in sensing devices.