EMAIL THIS PAGE TO A FRIEND
Marine drugs
Monacyclinones, New Angucyclinone Metabolites Isolated from Streptomyces sp. M7_15 Associated with the Puerto Rican Sponge Scopalina ruetzleri.
PMID 26230704
Abstract
During an investigation of new actinomycete species from Caribbean sponges for novel bioactive natural products, frigocyclinone (1), dimethyldehydrorabelomycin (3) and six new angucyclinone derivatives were isolated from Streptomyces sp. strain M7_15 associated with the sponge Scopalina ruetzleri. Of these, monacyclinones A-B (4-5) contain the core ring structure of dehydrorabelomycin (2) with the aminodeoxysugar found in frigocyclinone (1). Monacyclinone C (6) is a hydroxylated variant of frigocyclinone (1) and monacyclinone D (7) is a Baeyer Villiger derivative of (6) which also exists as the open chain hydrolysis product monacyclinone E (8). Monacyclinone F (9) contains two unique epoxide rings attached to the angucyclinone moiety and an additional aminodeoxysugar attached through an angular oxygen bond. All structures were confirmed through spectral analyses. Activity against rhabdomycosarcoma cancer cells (SJCRH30) after 48 h of treatment was observed with frigocyclinone (1; EC50 = 5.2 µM), monacyclinone C (6; 160 µM), monacyclinone E (8; 270 µM), and monacyclinone F (9; 0.73 µM). The strongest bioactivity against rhabdomycosarcoma cancer cells and gram-positive bacteria was exhibited by compound 9, suggesting that the extra aminodeoxysugar subunit is important for biological activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
194166
Methanol-d4, "100%", ≥99.96 atom % D, ≥99% (CP)
CD4O
721948 Methanol-d4, NMR reference standard, 99.8 atom % D, NMR tube size 5 mm × 8 in.
CD4O
721921 Methanol-d4, NMR reference standard, 99.8 atom % D, NMR tube size 3 mm × 8 in.
CD4O
444758
Methanol-d4, "100%", 99.96 atom % D
CD4O
423084
Methanol-d4, "100%", 99.96 atom % D
CD4O
535435
Methanol-d4, "100%", ≥99.96 atom % D, contains 0.03 % (v/v) TMS
CD4O
151947
Methanol-d4, ≥99.8 atom % D
CD4O
522023
Methanol-d4, 99.8 atom % D
CD4O
441384
Methanol-d4, ≥99.8 atom % D
CD4O
343803
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS
CD4O
530530
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS
CD4O
611646
Methanol-d4, ≥99.8 atom % D, contains 0.05 % (v/v) TMS
CD4O
439029
Methanol-d4, ≥99.8 atom % D, contains 0.1 % (v/v) TMS
CD4O
417653
Methanol-d4, ≥99.8 atom % D, contains 1 % (v/v) TMS
CD4O
422878
Methanol-d4, 99 atom % D
CD4O
569534
Methanol-d4, anhydrous, ≥99.8 atom % D
CD4O
570729
Methanol-d4, ≥99.8 atom % D, anhydrous
CD4O
177970
Pyridine-d5, "100%", ≥99.96 atom % D
C5D5N
437700
Pyridine-d5, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
C5D5N
532975
Pyridine-d5, ≥99.5 atom % D
C5D5N
532967
Pyridine-d5, ≥99.5 atom % D, contains 0.03 % (v/v) TMS
C5D5N
152323
Pyridine-d5, 99 atom % D
C5D5N