EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Metabolism of nitazoxanide in rats, pigs, and chickens: Application of liquid chromatography coupled to hybrid linear ion trap/Orbitrap mass spectrometer.
PMID 26231678
Abstract
Nitazoxanide (NTZ) is a nitrothiazole benzamide compound with a broad activity spectrum against parasites, Gram-positive and Gram-negative anaerobic bacteria, and viruses. In this study, hybrid linear ion trap/Orbitrap mass spectrometer providing a high mass resolution and accuracy was used to investigate the metabolism of NTZ in rats, pigs, and chickens. The results revealed that acetylation and glucuronidation were the main metabolic pathways in rats and pigs, whereas acetylation and sulfation were the major metabolic pathways in chickens, which indicated interspecies variations in drug metabolism and elimination. With the accurate mass data and the characteristic MS(n) product ions, we identified six metabolites in which tizoxanide and hydroxylated tizoxanide were phase I metabolites and tizoxanide glucuronide, tizoxanide glucose, tizoxanide sulfate and hydroxyl tizoxanide sulfate were phase II metabolites. Hydroxylated tizoxanide and tizoxanide glucose were identified for the first time. All the comprehensive data were provided to make out the metabolism of NTZ in rats, pigs and chickens more clearly.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N0290
Nitazoxanide, ≥98% (HPLC)
C12H9N3O5S