EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Two Schiff-base fluorescent sensors for selective sensing of aluminum (III): Experimental and computational studies.
PMID 26232579
Abstract
Two Schiff-base fluorescent sensors have been synthesized, which both can act as fluorescent probes for Al(3+), upon addition of Al(3+), they exhibit a large fluorescence enhancement which might be attributed to the formation of 1:1 ligand-Al complexes which inhibit photoinduced electron transfer (PET) progress, and that the proposed binding modes of the sensors and Al(3+) are identified by theoretical calculations.