EMAIL THIS PAGE TO A FRIEND
Nature communications
An apoptosis-enhancing drug overcomes platinum resistance in a tumour-initiating subpopulation of ovarian cancer.
PMID 26234182
Abstract
High-grade serous ovarian cancers (HGSCs) are deadly malignancies that relapse despite carboplatin chemotherapy. Here we show that 16 independent primary HGSC samples contain a CA125-negative population enriched for carboplatin-resistant cancer initiating cells. Transcriptome analysis reveals upregulation of homologous recombination DNA repair and anti-apoptotic signals in this population. While treatment with carboplatin enriches for CA125-negative cells, co-treatment with carboplatin and birinapant eliminates these cells in HGSCs expressing high levels of the inhibitor of apoptosis protein cIAP in the CA125-negative population. Birinapant sensitizes CA125-negative cells to carboplatin by mediating degradation of cIAP causing cleavage of caspase 8 and restoration of apoptosis. This co-therapy significantly improves disease-free survival in vivo compared with either therapy alone in tumour-bearing mice. These findings suggest that therapeutic strategies that target CA125-negative cells may be useful in the treatment of HGSC.