EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Convergent synthesis of 4,5-branched inner-core oligosaccharides of lipopoly- and lipooligosaccharides.
PMID 26235297
Abstract
The convergent synthesis of branched inner-core oligosaccharides of lipopoly- and lipooligosaccharide with a 3-deoxy-d-manno-oct-2-ulosonic acid (Kdo) disaccharide acceptor was achieved. The l-glycero-d-manno-heptopyranose (Hep) units for the branched core oligosaccharide Galβ(1-4)Glcβ(1-4)Hep and Hepα(1-3)Hep were prepared from the corresponding Hep building blocks. To obtain 4,5-branched core oligosaccharide structures, the common acceptor Kdoα(2-4)Kdo was glycosylated with the Hep units.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D110000
3,5-Dihydroxybenzoic acid, 97%
C7H6O4