EMAIL THIS PAGE TO A FRIEND
European Journal of Pharmaceutics and Biopharmaceutics
Bioadhesive vaginal drug delivery of nystatin using a derivatized polymer: Development and characterization.
PMID 26235393