EMAIL THIS PAGE TO A FRIEND
Journal of clinical biochemistry and nutrition
The effects of moderate exercise on secretory IgA production in mice depends on dietary carbohydrate intake.
PMID 26236100
Abstract
Secretory immunoglobulin A (sIgA) is produced from intestinal mucosa and is essential in preventing infection. We analyzed the influence of moderate exercise on intestinal sIgA production and antioxidative function under different carbohydrate nutritional conditions. Thirty-six mice were fed an experimental diet for 10 weeks-a high-carbohydrate (HC) diet, a low-carbohydrate (LC) diet, or a control (C) diet. After 1 week on the experimental diets, mice were divided into sedentary and exercise groups (n = 6/group), where the exercise consisted of treadmill running for 30 min/day at 11 m/min for 6 days/week in 9 consecutive weeks. Intestinal sIgA levels in the exercise groups fed C or LC diets were significantly lower compared with the parallel sedentary groups, or exercise-group mice fed HC diet. Expression of the polymeric immunoglobulin receptor (pIgR) in the small intestine was significantly higher in the exercise group fed a HC diet. Superoxide dismutase activity in the small intestine was higher in the exercise group than in the sedentary group, with no effects resulting from intake carbohydrate levels. Our results indicated that moderate exercise reduced the levels of intestinal sIgA depending on decreasing of carbohydrate intake, which is connected with the expression of pIgR.