EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Online 2D-LC-MS/MS assay to quantify therapeutic protein in human serum in the presence of pre-existing antidrug antibodies.
PMID 26237058
Abstract
The formation of antidrug antibodies (ADA) can interfere with the accurate quantitation of therapeutic proteins, leading to significantly underestimated drug concentrations and confounded pharmacokinetic (PK) data interpretation. Although highly desirable, development of ADA-tolerant bioanalytical methods enabling unbiased measurement of both free and ADA-bound drug presents a considerable challenge. We report herein the development and validation of a robust LC-MS assay capable of quantifying therapeutic protein immunoglobulin A1 protease (IgAP) in human serum in the presence of pre-existing anti-IgAP antibodies. The procedure included sodium dodecyl sulfate (SDS) denaturation and chemical reduction of serum proteins to dissociate ADA-drug bindings, followed by tryptic digestion of protein pellets and subsequent LC-MS analysis of the surrogate IgAP peptide using stable isotope labeled peptide internal standard. Substantial enhancements in the sensitivity and selectivity were achieved by the combination of online two-dimensional reversed-phase LC (2D-LC) operated in high and low pH buffers, respectively, for efficient enrichment and quantitation of the surrogate peptide by multiple-reaction monitoring (MRM) mass spectrometry. Unlike ligand-binding assay, our method is not prone to interferences from ADA, allowing accurate and precise measurement of the IgAP in the range of 0.05 to 10 μg/mL in 25 μL of human serum with a wide range of anti-IgAP antibody levels. The intra- and inter-run precision (coefficient of variation (CV%)) was within 11.5% and 10.5%, respectively, and the bias was within ±7.1% for all quality control (QC) concentrations. With little modification, the described method can readily be applicable to the quantitation of other biotherapeutic proteins in the ADA-positive clinical matrices.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2886
3-Indoleacetic acid, plant cell culture tested, crystalline
C10H9NO2
I3750
3-Indoleacetic acid, 98%
C10H9NO2
W248703
Formic acid, ≥95%, FCC, FG
CH2O2
I5148
Indole-3-acetic acid sodium salt, BioReagent, plant cell culture tested, ≥98%
C10H8NNaO2
I1149
Iodoacetamide, BioUltra
C2H4INO
I6125
Iodoacetamide, ≥99% (NMR), crystalline
C2H4INO
A3221
Iodoacetamide, Single use vial of 56 mg
C2H4INO
I5161
Iodoacetamide, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production
C2H4INO
68957
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, for electrophoresis, SDS-PAGE tested
C9H15O6P · HCl
C4706
Tris(2-carboxyethyl)phosphine hydrochloride, powder, ≥98%
C9H15O6P · HCl
75259
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, ≥98% (NMR)
C9H15O6P · HCl
646547
Tris(2-carboxyethyl)phosphine hydrochloride solution, 0.5 M, pH 7.0(aqueous solution; pH was adjusted with ammonium hydroxide)
C9H15O6P · HCl
U0631
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
CH4N2O
U5378
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
CH4N2O
U4883
Urea, 8 M (after reconstitution with 16 mL high purity water)
CH4N2O
U6504
Urea, for electrophoresis
CH4N2O
U4884
Urea, meets USP testing specifications
CH4N2O
U5128
Urea, ACS reagent, 99.0-100.5%
CH4N2O
U1250
Urea, ReagentPlus®, ≥99.5%, pellets
CH4N2O
51456
Urea, BioUltra, for molecular biology, ≥99.5% (T)
CH4N2O
33247
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
CH4N2O
15604
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
CH4N2O
U7129
Urea solution, 40 % (w/v) in H2O
CH4N2O
51457
Urea solution, BioUltra, ~8 M in H2O
CH4N2O