EMAIL THIS PAGE TO A FRIEND
Journal of traditional Chinese medicine = Chung i tsa chih ying wen pan
Heat induces adenosine triphosphate release from mast cells in vitro: a putative mechanism for moxibustion.
PMID 26237838
Abstract
To investigate the role of adenosine tri-phosphate (ATP) purinergic signaling in mast cells (MCs) modulated by heat to further understand the molecular mechanisms of moxibustion. Skin temperatures induced by monkshood cake moxibustion were evaluated by measuring the Neiguan acupoint (PC 6) from 31 participants with a digital thermocouple thermometer. Temperatures of 43 °C and 52 °C were applied to cultured human leukemia mast cell line HMC-1 in vitro. Calcium fluorescence was applied to detect intracellular Ca2+ ([Ca2+]). Extracellular ATP contents were measured by luciferin-luciferase assay. Maximum skin temperatures mostly ranged from 40-45 °C , but some reached up to 50 °C. Both 43 °C and 52 °C induced MC degranulation, which was accompanied by an increase in [Ca2+] and ATP release. Complexing extracellular Ca2+ with 5 mM ethylene glycol-bis (β-aminoethyl ether)-N,N,N',N'-tetraacetic acid (EGTA) inhibited the noxious, heat-induced elevation of [Ca2+]i and prevented the enhanced ATP secretion by those. cells at 52 °C, but not 43 °C. Monkshood cake moxibustion can generate heat sufficient to trigger cellular events of MCs, including degranulation, [Ca2+]i elevation, and ATP release, suggesting that purinergic signals originating from MCs are possibly the initiating response of acupoints to moxibustion.