EMAIL THIS PAGE TO A FRIEND
Oncology reports
Fusion of cancer stem cells and mesenchymal stem cells contributes to glioma neovascularization.
PMID 26238144
Abstract
The ability of tumor cells to autonomously generate tumor vessels has received considerable attention in recent years. However, the degree of autonomy is relative. Meanwhile, the effect of bone marrow-derived mesenchymal stem cells (BMSCs) on tumor neovascularization has not been fully elucidated. The present study aimed to illuminate whether cell fusion between glioma stem cells and BMSC is involved in glioma neovascularization. BMSCs were isolated from transgenic nude mice, of which all nucleated cells express green fluorescent protein (GFP). The immunophenotype and multilineage differentiation potential of BMSC were confirmed. SU3 glioma stem/progenitor cells were transfected with red fluorescent protein (SU3-RFP cells). In a co-culture system of BMSC-GFP and SU3-RFP, RFP+/GFP+ cells were detected and isolated by dual colors using FACS. The angiogenic effect of RFP+/GFP+ cells was determined in vivo and in vitro. Flow cytometry analysis showed that BMSC expressed high levels of CD105, C44, and very low levels of CD45 and CD11b. When co-cultured with SU3-RFP, 73.8% of cells co-expressing RFP and GFP were identified as fused cells in the 5th generation. The fused cells exhibited tube formation ability in vitro and could give rise to a solid tumor and form tumor blood vessels in vivo. In the dual-color orthotopic model of transplantable xenograft glioma, yellow vessel-like structures that expressed CD105, RFP and GFP were identified as de novo-formed vessels derived from the fused cells. The yellow vessels observed in the tumor-bearing mice directly arose from the fusion of BMSCs and SU3-RFP cells. Thus, cell fusion is one of the driving factors for tumor neovascularization.