EMAIL THIS PAGE TO A FRIEND
Obesity (Silver Spring, Md.)
Hepatic and visceral adipose tissue 11βHSD1 expressions are markers of body weight loss after bariatric surgery.
PMID 26239572
Abstract
Cortisolemia and 11βHSD1 in liver and adipose tissue are altered in obesity. However, their participation in the development of obesity remains unclear. This study analyzed these parameters in the transition from morbid to type 1 obesity after bariatric surgery. A group of 34 patients with morbid obesity and 22 nonobese subjects were recruited. Initial hypothalamus-pituitary-adrenal (HPA) basal activity and 11βHSD1 mRNA expression in liver, subcutaneous (SAT), and visceral adipose tissue (VAT) were evaluated. A year after bariatric surgery (weight loss of 48 kg), these parameters were reappraised in plasma, SAT, and liver. Body weight loss was accompanied by a downshift in basal HPA activity and 11βHSD1 expression in SAT. In patients with morbid obesity, 11βHSD1 expression correlated positively with BMI in VAT and negatively in liver at 6 and 12 months after surgery. In SAT, a correlation was observed with body weight only when patients showed type 1 obesity. Insulin, glucose, and HOMA correlated positively with all the HPA indicators and 11βHSD1 expression in SAT. Body weight loss after bariatric surgery is accompanied by a downshift in basal HPA activity. Hepatic and VAT 11βHSD1 expressions in morbid obesity are predictors of body weight loss.