EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Synergetic deoxy reforming of cellulose and fatty acid esters for liquid hydrocarbon-rich oils.
PMID 26241841
Abstract
A series of liquid hydrocarbons (alkylbenzenes, alkanes, and alkenes) were obtained by a synergetic deoxy reforming (SDR) process of cellulose and linoleic acid methyl ester (LAME) at 350°C and 4-6MPa in a closed system without external source of hydrogen. The liquid product was obtained with a yield of 15wt% at a LAME/cellulose ratio of 0.2. In contrast, the direct deoxy reforming of cellulose produces oil that contains plenty of phenols and oxygen-containing compounds. Due to the insufficiency of water employed (30wt%), a radical reaction pathway was proposed. Quantum chemical calculations indicate that the radicals from LAME interfere with the reactions of the intermediate products from cellulose, being responsible for the removal of phenols and the formation of hydrocarbons. The SDR process offers an embryonic insight in an alternative technique for preparation of hydrocarbon fuels.