EMAIL THIS PAGE TO A FRIEND
AMB Express
Production of a modified peptide clavanin in Pichia pastoris: cloning, expression, purification and in vitro activities.
PMID 26243173
Abstract
Antimicrobial peptides are one of the most promising peptide-based drugs due to their enormous potential as novel biopharmaceuticals in both human and animal industries. In order to develop strategies to over produce such molecules, heterologous production of a modified version of clavanin A, here named clavanin MO (clavMO), was successfully achieved in the methylothopic yeast Pichia pastoris. ClavMO was fused to thioredoxin as a carrier protein and the construction was tested using two promoters, PAOX1 and PGAP, based on either induced or constitutive expression systems, respectively. After growth in 5 L Bioreactor, clavMO-thio was recovered and purified through size exclusion chromatography. Our findings show that both constitutive and inducible expression systems produce active clavMO fused to thioredoxin against both Gram-negative Klebsiella pneumoniae and Gram-positive Staphylococcus aureus microorganisms.