EMAIL THIS PAGE TO A FRIEND
Nature communications
Mto2 multisite phosphorylation inactivates non-spindle microtubule nucleation complexes during mitosis.
PMID 26243668
Abstract
Microtubule nucleation is highly regulated during the eukaryotic cell cycle, but the underlying molecular mechanisms are largely unknown. During mitosis in fission yeast Schizosaccharomyces pombe, cytoplasmic microtubule nucleation ceases simultaneously with intranuclear mitotic spindle assembly. Cytoplasmic nucleation depends on the Mto1/2 complex, which binds and activates the γ-tubulin complex and also recruits the γ-tubulin complex to both centrosomal (spindle pole body) and non-centrosomal sites. Here we show that the Mto1/2 complex disassembles during mitosis, coincident with hyperphosphorylation of Mto2 protein. By mapping and mutating multiple Mto2 phosphorylation sites, we generate mto2-phosphomutant strains with enhanced Mto1/2 complex stability, interaction with the γ-tubulin complex and microtubule nucleation activity. A mutant with 24 phosphorylation sites mutated to alanine, mto2[24A], retains interphase-like behaviour even in mitotic cells. This provides a molecular-level understanding of how phosphorylation 'switches off' microtubule nucleation complexes during the cell cycle and, more broadly, illuminates mechanisms regulating non-centrosomal microtubule nucleation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3553
Acrylamide, for electrophoresis, ≥99% (HPLC), powder
C3H5NO
A8887
Acrylamide, suitable for electrophoresis, ≥99%
C3H5NO
A9099
Acrylamide, for molecular biology, ≥99% (HPLC)
C3H5NO
A4058
Acrylamide solution, 40%, for electrophoresis, sterile-filtered
C3H5NO
A5508
Adenosine 5′-phosphosulfate sodium salt, ≥85%
C10H14N5O10PS
A3678
Ammonium persulfate, for molecular biology, for electrophoresis, ≥98%
H8N2O8S2
A9164
Ammonium persulfate, for molecular biology, powder, ≥98%
H8N2O8S2
A7460
Ammonium persulfate, BioXtra, ≥98.0%
H8N2O8S2
09913
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
H8N2O8S2
11020
DL-Arginine, ≥97.0% (NT)
C6H14N4O2
RDD001
Guanidine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99%
CH5N3 · HCl
PHG0006
Guanidine hydrochloride, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production.
CH5N3 · HCl
G4505
Guanidine hydrochloride, ≥99% (titration), organic base and chaeotropic agent
CH5N3 · HCl
50933
Guanidine hydrochloride, BioUltra, for molecular biology, ≥99.5% (AT)
CH5N3 · HCl
G3272
Guanidine hydrochloride, for molecular biology, ≥99%
CH5N3 · HCl
G7153
Guanidine hydrochloride, BioXtra, ≥99.5% (titration)
CH5N3 · HCl
G4630
Guanidine hydrochloride, ≥98%
CH5N3 · HCl
50940
Guanidine hydrochloride, ≥99.0% (AT)
CH5N3 · HCl
50950
Guanidine hydrochloride, ≥98%
CH5N3 · HCl
G9284
Guanidine hydrochloride solution, 8 M in H2O (filtered)
CH5N3 · HCl
50937
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
CH5N3 · HCl
792527
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
C3H4N2
I5513
Imidazole, for molecular biology, ≥99% (titration)
C3H4N2
I0250
Imidazole, ≥99% (titration), crystalline
C3H4N2
I2399
Imidazole, ACS reagent, ≥99% (titration)
C3H4N2
56750
Imidazole, puriss. p.a., ≥99.5% (GC)
C3H4N2
I202
Imidazole, ReagentPlus®, 99%
C3H4N2
56748
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
C3H4N2
56749
Imidazole, BioUltra, ≥99.5% (GC)
C3H4N2
68268
Imidazole buffer Solution, BioUltra, 1 M in H2O
C3H4N2