EMAIL THIS PAGE TO A FRIEND
Behavioral ecology and sociobiology
Conspecific recognition and aggression reduction to familiars in newly weaned, socially plastic mammals.
PMID 26246656
Abstract
Recognising conspecifics and behaving appropriately towards them is a crucial ability for many species. Grey seals (Halichoerus grypus) show varying capabilities in this regard: mother-pup recognition has been demonstrated in some geographical populations but is absent in others, yet there is evidence that individuals aggregate with prior associates. The recognition capabilities of newly weaned grey seal pups were investigated using class recognition trials within the habituation/dishabituation paradigm. Trials took place in pens, using pairs of individuals that either had previously cohabited (familiar) or that had never met before (stranger). Frequencies of olfactory and visual investigative behaviours ('checks') and aggressive interactions were recorded during trials. Familiar individuals recognised each other: paired strangers showed significantly more checks and aggressive interactions than were seen in trials pairing familiars. Oxytocin concentrations in post-trial plasma samples were analysed to investigate the underlying physiology modulating recognition abilities; however, no significant differences were detected between familiar or stranger trials. This study demonstrates that at a young age, grey seals can recognise individuals they have previously encountered. Recognition abilities in this species have adaptive value by allowing the reduction of costly aggressive interactions between familiar conspecifics, which is often cited as the first step towards the evolution of sociality in a species. This study is the first with wild subjects to find conspecific recognition abilities in a pinniped species outside of reproductive contexts. It demonstrates that even largely solitary species can be capable of recognition and pro-social behaviours that benefit them during times when they must aggregate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O3251
Oxytocin
C43H66N12O12S2
O4375
Oxytocin, lyophilized powder, ~15 IU/mg solid (Prepared from synthetic oxytocin)
C43H66N12O12S2