EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Single-Layer Light-Emitting Diodes Using Organometal Halide Perovskite/Poly(ethylene oxide) Composite Thin Films.
PMID 26247326
Abstract
Organometal halide perovskite and poly(ethylene oxide) composite thin films are studied. Single-layer light-emitting diodes using the composite thin film sandwiched between indium tin oxide and indium-gallium eutectic alloy exhibit a low turn-on voltage and high brightness because of the ionic conductivity of the composite film and the formation of a p-i-n homojunction.