EMAIL THIS PAGE TO A FRIEND
Drug design, development and therapy
A novel in situ gel base of deacetylase gellan gum for sustained ophthalmic drug delivery of ketotifen: in vitro and in vivo evaluation.
PMID 26251573
Abstract
In this study, an ion-activated ketotifen ophthalmic delivery system was developed by using a natural polysaccharide, deacetylase gellan gum. Its rheological characteristics, stability, in vitro gelation, release in vitro, and pharmacodynamic activity in vivo were investigated. The formulation had an optimum viscosity that will allow easy drop as a liquid, which then underwent a rapid sol-gel transition due to ionic interaction. There were negligible alterations in the initial values of viscosity of the formulations over a storage period of 180 days. The in vitro release profiles indicated that the release of ketotifen from in situ gels exhibited a sustained feature. Scintigraphic studies indicated that deacetylase gellan gum could increase the residence time of the formulation. At the same dose, in situ gels demonstrated a typical sustained and prolonged drug-effects behavior compared with the common drops.