PloS one
Bortezomib Plus Continuous B Cell Depletion Results in Sustained Plasma Cell Depletion and Amelioration of Lupus Nephritis in NZB/W F1 Mice.
PMID 26252021
Abstract
NZB/W F1 mice were treated with: 1) anti-CD20, 2) anti-CD20 plus bortezomib, 3) anti-CD20 plus anti-LFA-1/anti-VLA-4 blocking antibodies, 4) anti-CD20 plus bortezomib and anti-LFA-1/anti-VLA4 blocking antibodies. Short- and long-lived plasma cells including autoreactive cells in the bone marrow and spleen were enumerated by flow cytometry and ELISPOT seven days after treatment. Based on these data in another experiment, mice received one cycle of anti-CD20 plus bortezomib followed by four cycles of anti-CD20 therapy every 10 days and were monitored for its effect on plasma cells and disease. Short-lived plasma cells in bone marrow and spleen were efficiently depleted by all regimens targeting plasma cells. Conversely, LLPCs and anti-dsDNA-secreting plasma cells in bone marrow and spleen showed resistance to depletion and were strongly reduced by bortezomib plus anti-CD20. The effective depletion of plasma cells by bortezomib complemented by the continuous depletion of their precursor B cells using anti-CD20 promoted the persistent reduction of IgG anti-dsDNA antibodies, delayed nephritis and prolonged survival in NZB/W F1 mice. These findings suggest that the effective depletion of LLPCs using bortezomib in combination with a therapy that continuously targeting B cells as their precursors may prevent the regeneration of autoreactive LLPCs and, thus, might represent a promising treatment strategy for SLE and other (auto)antibody-mediated diseases.