EMAIL THIS PAGE TO A FRIEND
Journal of Toxicology and Environmental Health. Part A
4-Hydroxynonenal: A Superior Oxidative Biomarker Compared to Malondialdehyde and Carbonyl Content Induced by Carbon Tetrachloride in Rats.
PMID 26252470