EMAIL THIS PAGE TO A FRIEND
Nucleosides, nucleotides & nucleic acids
Nucleoside Analog Activity in Malignant Melanoma Cell Lines.
PMID 26252632
Abstract
Mitochondrial deoxyguanosine kinase (dGK), is an enzyme responsible for activation of nucleoside analogs (NAs) to phosphorylated compounds which exert profound cytotoxicity, especially in hematological malignancies. Screening malignant melanoma cell lines against NAs revealed high sensitivity to several of them. This was believed to be due to the high levels of dGK expression in these cells. Downregulation of dGK in the melanoma cell line RaH5 using siRNA did not cause resistance to NAs as expected, but instead cells became more sensitive. This was probably partly due to the increased activity of another mitochondrial enzyme, thymidine kinase 2, seen in transfected cells.