EMAIL THIS PAGE TO A FRIEND
Genes & development
AUF1 promotes let-7b loading on Argonaute 2.
PMID 26253535
Abstract
Eukaryotic gene expression is tightly regulated post-transcriptionally by RNA-binding proteins (RBPs) and microRNAs. The RBP AU-rich-binding factor 1 (AUF1) isoform p37 was found to have high affinity for the microRNA let-7b in vitro (Kd = ∼ 6 nM) in cells. Ribonucleoprotein immunoprecipitation, in vitro association, and single-molecule-binding analyses revealed that AUF1 promoted let-7b loading onto Argonaute 2 (AGO2), the catalytic component of the RNA-induced silencing complex (RISC). In turn, AGO2-let-7 triggered target mRNA decay. Our findings uncover a novel mechanism by which AUF1 binding and transfer of microRNA let-7 to AGO2 facilitates let-7-elicited gene silencing.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EMU056441
MISSION® esiRNA, targeting mouse Eif2c2