EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
MAZR and Runx Factors Synergistically Repress ThPOK during CD8+ T Cell Lineage Development.
PMID 26254341
Abstract
Th-inducing Pox virus and zinc finger/Krüppel-like factor (ThPOK) is a key commitment factor for CD4(+) lineage T cells and is essential for the maintenance of CD4 lineage integrity; thus, the expression of ThPOK has to be tightly controlled. In this article, we demonstrate that Myc-associated zinc finger-related factor (MAZR) and Runt-related transcription factor 1 (Runx1) together repressed ThPOK in preselection double-positive thymocytes, whereas MAZR acted in synergy with Runx3 in the repression of ThPOK in CD8(+) T cells. Furthermore, MAZR-Runx1 and MAZR-Runx3 double-mutant mice showed enhanced derepression of Cd4 in double-negative thymocytes and in CD8(+) T cells in comparison with Runx1 or Runx3 single-deficient mice, respectively, indicating that MAZR modulates Cd4 silencing. Thus, our data demonstrate developmental stage-specific synergistic activities between MAZR and Runx/core-binding factor β (CBFβ) complexes. Finally, retroviral Cre-mediated conditional deletion of MAZR in peripheral CD8(+) T cells led to the derepression of ThPOK, thus showing that MAZR is also part of the molecular machinery that maintains a repressed state of ThPOK in CD8(+) T cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H9268
Hexadimethrine bromide, ≥94% (titration)