EMAIL THIS PAGE TO A FRIEND
RSC advances
Small lipopeptides possess anti-biofilm capability comparable to daptomycin and vancomycin.
PMID 26257894
Abstract
Antibiotic resistance, to a large extent, is related to the formation of bacterial biofilms. Thus, compounds with anti-biofilm capability are of practical importance. Inspired by the recent discovery of two amino acid lipopeptides from marine bacteria, we constructed a family of small lipopeptides with 2-3 amino acids. While no antimicrobial activity was found for anionic lipopeptides, cationic candidates are potent against Staphylococcus strains, such as methicillin-resistant