EMAIL THIS PAGE TO A FRIEND
The Journal of experimental medicine
T cell metabolism drives immunity.
PMID 26261266
Abstract
Lymphocytes must adapt to a wide array of environmental stressors as part of their normal development, during which they undergo a dramatic metabolic remodeling process. Research in this area has yielded surprising findings on the roles of diverse metabolic pathways and metabolites, which have been found to regulate lymphocyte signaling and influence differentiation, function and fate. In this review, we integrate the latest findings in the field to provide an up-to-date resource on lymphocyte metabolism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
94640
DL-Valine, ReagentPlus®, ≥99.0% (NT)
C5H11NO2
W344400
DL-Valine, ≥97%
C5H11NO2