EMAIL THIS PAGE TO A FRIEND
International journal of clinical and experimental pathology
Nigella sativa amliorates inflammation and demyelination in the experimental autoimmune encephalomyelitis-induced Wistar rats.
PMID 26261504
Abstract
Multiple sclerosis (MS) is the major, immune-mediated, demyelinating neurodegenerative disease of the central nervous system (CNS). Experimental autoimmune encephalomyelitis (EAE) is a well-established animal model of MS. The aim of the present study was to investigate the protective and ameliorative effects of N. sativa seeds (2.8 g/kg body weight) in EAE-induced Wistar rats. EAE-induced rats were divided into: 1- EAE-induced rats ("EAE" group). 2- "N. sativa + EAE" group received daily oral administration of N. sativa 2 weeks prior EAE induction until the end of the experiment. 3- "EAE + N. sativa" group received daily oral administration of N. sativa after the appearance of first clinical signs until the end of the experiment. All animals were decapitated at the 28th day post EAE-induction. EAE was investigated using histopathological, immunohistochemical and ultrastructural examinations in addition to determination of some oxidative stress parameters in the cerebellum and medulla. N. sativa suppressed inflammation observed in EAE-induced rats. In addition, N. sativa enhanced remyelination in the cerebellum. Moreover, N. sativa reduced the expression of transforming growth factor beta 1 (TGF β1). N. sativa seeds could provide a promising agent effective in both the protection and treatment of EAE.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
302066
1,2-Dinitrobenzene, ≥99%
C6H4N2O4
126632
1,2-Dinitrobenzene, 97%
C6H4N2O4