EMAIL THIS PAGE TO A FRIEND
International journal of clinical and experimental pathology
rhEPO affects apoptosis in hippocampus of aging rats by upregulating SIRT1.
PMID 26261574
Abstract
The aim of this study was to elucidate the signaling pathway involved in the anti-aging effect of erythropoietin (EPO) and to clarify whether recombinant human EPO (rhEPO) affects apoptosis in the aging rat hippocampus by upregulating Sirtuin 1 (SIRT1). In this study, a rat model of aging was established using D-galactose. Behavioral changes were monitored by the Morris water maze test. Using immunohistochemistry, we studied the expression of SIRT1, B-cell lymphoma/leukemia-2 gene (Bcl-2), and Bcl-2 associated X protein (Bax) expression, and apoptotic cells in the hippocampus of a rat model of aging in which rhEPO was intraperitoneally injected. The escape latency in rats from the EPO group shortened significantly; however, the number of platform passes increased significantly from that in the D-gal group (P < 0.05). Compared to the D-gal group, in the EPO group, the number of SIRT1 and Bcl-2-positive cells increased (P < 0.05), but the number of Bax-positive cells and apoptotic cells decreased in the hippocampus of aging rats (P < 0.05). These results suggest that rhEPO regulates apoptosis-related genes and affects apoptosis in the hippocampus of aging rats by upregulating SIRT. This may be one of the important pathways underlying the anti-aging property of EPO.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SRP6158
EPO human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)