EMAIL THIS PAGE TO A FRIEND
International Journal of Food Microbiology
Evaluation of different buffered peptone water (BPW) based enrichment broths for detection of Gram-negative foodborne pathogens from various food matrices.
PMID 26267889