EMAIL THIS PAGE TO A FRIEND
Investigational New Drugs
Chloroquine inhibits the malignant phenotype of glioblastoma partially by suppressing TGF-beta.
PMID 26271735
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6628
Chloroquine diphosphate salt, solid, ≥98%
C18H26ClN3 · 2H3PO4