EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Two phosphonium ionic liquids with high Li(+) transport number.
PMID 26272339
Abstract
This work presents the physicochemical characterization of two ionic liquids (ILs) with small phosphonium cations, triethylpenthylphosphonium bis(trifluoromethanesulfonyl)imide ([P2225][Tf2N]) and (2-methoxyethyl)trimethylphosphonium bis(trifluoromethanesulfonyl)imide ([P222(201)][Tf2N]), and their mixtures with Li(+). Properties such as the electrochemical window, density, viscosity and ionic conductivity are presented. The diffusion coefficient was obtained using two different techniques, PGSE-NMR and Li electrodeposition with microelectrodes. In addition, the Li(+) transport number was calculated using the PGSE-NMR technique and an electrochemical approach. The use of these three techniques showed that the PGSE-NMR technique underestimates the diffusion coefficient for charged species. The Li(+) transport number was found to be as high as 0.54. Raman spectroscopy and molecular dynamics simulations were used to evaluate the short-range structure of the liquids. These experiments suggested that the interaction between the Li(+) and the Tf2N(-) anion is similar to that seen with other ILs containing the same anion. However, the MD simulations also showed that the Li(+) ions interact differently with the cation containing an alkyl ether chain. The results found in this work suggest that these Li(+) mixtures have promising potential to be applied as electrolytes in batteries.
Related Materials