EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Heart and circulatory physiology
Dynamics of enhanced mitochondrial respiration in female compared with male rat cerebral arteries.
PMID 26276815
Abstract
Mitochondrial respiration has never been directly examined in intact cerebral arteries. We tested the hypothesis that mitochondrial energetics of large cerebral arteries ex vivo are sex dependent. The Seahorse XFe24 analyzer was used to examine mitochondrial respiration in isolated cerebral arteries from adult male and female Sprague-Dawley rats. We examined the role of nitric oxide (NO) on mitochondrial respiration under basal conditions, using N(ω)-nitro-l-arginine methyl ester, and following pharmacological challenge using diazoxide (DZ), and also determined levels of mitochondrial and nonmitochondrial proteins using Western blot, and vascular diameter responses to DZ. The components of mitochondrial respiration including basal respiration, ATP production, proton leak, maximal respiration, and spare respiratory capacity were elevated in females compared with males, but increased in both male and female arteries in the presence of the NOS inhibitor. Although acute DZ treatment had little effect on mitochondrial respiration of male arteries, it decreased the respiration in female arteries. Levels of mitochondrial proteins in Complexes I-V and the voltage-dependent anion channel protein were elevated in female compared with male cerebral arteries. The DZ-induced vasodilation was greater in females than in males. Our findings show that substantial sex differences in mitochondrial respiratory dynamics exist in large cerebral arteries and may provide the mechanistic basis for observations that the female cerebral vasculature is more adaptable after injury.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
266167
Manganese, chips, thickness <2.0 mm, 99%
Mn
266132
Manganese, powder, −325 mesh, ≥99% trace metals basis
Mn
463728
Manganese, powder, ≥99.9% trace metals basis
Mn
GF74177736
Manganese, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 98.7%
Mn
GF99005193
Manganese, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 98.7%
Mn
GF90559794
Manganese, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 98.7%
Mn
GF03428625
Manganese, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 98.7%
Mn
GF31519975
Manganese, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 98.7%
Mn
GF60677646
Manganese, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 98.7%
Mn
GF64815239
Manganese, foil, not light tested, 50x50mm, thickness 0.002mm, permanent polyester support, 98.7%
Mn
GF41418378
Manganese, foil, not light tested, 50x50mm, thickness 0.003mm, permanent polyester support, 98.7%
Mn
GF33640860
Manganese, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 98.7%
Mn
GF85367605
Manganese, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 98.7%
Mn
GF87981564
Manganese, foil, not light tested, 50x50mm, thickness 0.006mm, permanent polyester support, 98.7%
Mn
GF53022833
Manganese, foil, not light tested, 50x50mm, thickness 0.007mm, permanent polyester support, 98.7%
Mn
GF71274258
Manganese, foil, not light tested, 50x50mm, thickness 0.008mm, permanent polyester support, 98.7%
Mn
GF70055524
Manganese, foil, not light tested, 50x50mm, thickness 0.009mm, permanent polyester support, 98.7%
Mn
GF86610539
Manganese, foil, not light tested, 50x50mm, thickness 0.0125mm, permanent polyester support, 98.7%
Mn
GF00832081
Manganese, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 98.7%
Mn
GF61911288
Manganese, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 98.7%
Mn
GF94645233
Manganese, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 98.7%
Mn
GF05710430
Manganese, foil, not light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 98.7%
Mn
GF27632055
Manganese, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 98.7%
Mn
GF11021094
Manganese, rod, 10mm, diameter 4.0mm, cast, 99.5%
Mn
GF34573697
Manganese, rod, 35mm, diameter 2.0mm, cast, 99.5%
Mn
GF38495885
Manganese, rod, 50mm, diameter 4.0mm, cast, 99.5%
Mn
GF42039918
Manganese, rod, 50mm, diameter 6.35mm, cast, 99.5%
Mn
GF83932768
Manganese, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
Mn
GF87355334
Manganese, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
Mn
GF15798730
Manganese, foil, not light tested, 100x100mm, thickness 0.0125mm, permanent polyester support, 98.7%
Mn