EMAIL THIS PAGE TO A FRIEND
Neuroimmunomodulation
All-trans-Retinoic Acid Modulates Nitric Oxide and Interleukin-17A Production by Peripheral Blood Mononuclear Cells from Patients with Alzheimer's Disease.
PMID 26278415