EMAIL THIS PAGE TO A FRIEND
Journal of Evolutionary Biology
Little evidence for morphological change in a resilient endemic species following the introduction of a novel predator.
PMID 26278629
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9635
DL-p-Chloroamphetamine hydrochloride, ≥94% (HPLC)
C9H12ClN · HCl