EMAIL THIS PAGE TO A FRIEND
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy
Ab initio and density functional theory calculations of molecular structure and vibrational spectra of 4-(2-Hydroxyethyl) piperazine-1-ethanesulfonic acid.
PMID 26278882