EMAIL THIS PAGE TO A FRIEND
Journal of anatomy
Musculoskeletal anatomy and feeding performance ofxa0pre-feeding engyodontic larvae of the European eelxa0(Anguilla anguilla).
PMID 26278932
Abstract
Being part of the elopomorph group of fishes, Anguillidae species show a leptocephalus larval stage. However, due to largely unknown spawning locations and habitats of their earliest life stages, as well as their transparency, these Anguilla larvae are rarely encountered in nature. Therefore, information regarding the early life history of these larvae, including their exogenous feeding strategy and feeding performance, is rather scarce. To better understand the structural basis and functional performance of larval feeding in captivity, the functional morphology of the cranial musculoskeletal system in pre- and first-feeding engyodontic leptocephali of the European eel (Anguilla anguilla) was studied. A 3D reconstruction of the feeding apparatus (head of the leptocephali < 1 mm) was used to visualize and describe the musculoskeletal changes throughout these stages. To analyze the ontogenetic changes in the functionality of the feeding apparatus towards the active feeding phase, 3D data of joints, levers and muscles derived from the reconstructions were used to estimate bite and joint reaction forces (JRFs). Observing a maximum estimated bite force of about 65 μN (and corresponding JRFs of 260 μN), it can be hypothesized that leptocephalus larvae are functionally constrained to feed only on soft food particles. Additionally, potential prey items are size delimited, based on the theoretically estimated average gape of these larvae of about 100 μm. This hypothesis appears to be in line with recent observations of a diet consisting of small and/or gelatinous prey items (Hydrozoa, Thaliacea, Ctenophora, Polycystenia) found in the guts of euryodontic leptocephalus larvae.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
443271
Dodecenylsuccinic anhydride, mixture of isomers, technical grade, 90%
C16H26O3