EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Metabolic fate of endogenous molecular damage: Urinary glutathione conjugates of DNA-derived base propenals as markers of inflammation.
PMID 26283391
Abstract
Although mechanistically linked to disease, cellular molecules damaged by endogenous processes have not emerged as significant biomarkers of inflammation and disease risk, due in part to poor understanding of their pharmacokinetic fate from tissue to excretion. Here, we use systematic metabolite profiling to define the fate of a common DNA oxidation product, base propenals, to discover such a biomarker. Based on known chemical reactivity and metabolism in liver cell extracts, 15 candidate metabolites were identified for liquid chromatography-coupled tandem mass spectrometry (LC-MS/MS) quantification in urine and bile of rats treated with thymine propenal (Tp). Analysis of urine revealed three metabolites (6% of Tp dose): thymine propenoate and two mercapturate derivatives of glutathione conjugates. Bile contained an additional four metabolites (22% of Tp dose): cysteinylglycine and cysteine derivatives of glutathione adducts. A bis-mercapturate was observed in urine of untreated rats and increased approximately three- to fourfold following CCl4-induced oxidative stress or treatment with the DNA-cleaving antitumor agent, bleomycin. Systematic metabolite profiling thus provides evidence for a metabolized DNA damage product as a candidate biomarker of inflammation and oxidative stress in humans.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
110221
Acrolein, contains hydroquinone as stabilizer, 90%
C3H4O
CDS019917
Adenine phosphate salt, AldrichCPR
C5H5N5 · H3O4P
W248703
Formic acid, ≥95%, FCC, FG
CH2O2
G4251
L-Glutathione reduced, ≥98.0%
C10H17N3O6S
G6529
L-Glutathione reduced, BioXtra, ≥98.0%
C10H17N3O6S
G6013
L-Glutathione reduced, cell culture tested, BioReagent, ≥98.0%, powder
C10H17N3O6S
P2222
Potassium phosphate dibasic, meets USP testing specifications
HK2O4P
450200
Potassium phosphate dibasic, 99.95% trace metals basis
HK2O4P
P8281
Potassium phosphate dibasic, reagent grade, ≥98.0%
HK2O4P
60353
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
HK2O4P
P8584
Potassium phosphate dibasic solution, 1.0 M
HK2O4P
P5655
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
H2KO4P
RES20760-A7
Potassium phosphate monobasic, PharmaGrade, NF, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
H2KO4P
P8416
Potassium phosphate monobasic, plant cell culture tested, ≥99%
H2KO4P
P9791
Potassium phosphate monobasic, for molecular biology, ≥98.0%
H2KO4P
229806
Potassium phosphate monobasic, 99.99% trace metals basis
H2KO4P
P5379
Potassium phosphate monobasic, ReagentPlus®
H2KO4P
60218
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
H2KO4P
30-4310
Trifluoroacetic acid, SAJ special grade, ≥99.0%
C2HF3O2
91709
Trifluoroacetic acid solution, BioReagent, suitable for protein sequencing, ~25% in H2O
C2HO2F3