EMAIL THIS PAGE TO A FRIEND
BMC complementary and alternative medicine
Antioxidant potential, total phenolic and total flavonoid contents of Rhododendron anthopogonoides and its protective effect on hypoxia-induced injury in PC12 cells.
PMID 26283543
Abstract
Rhododendron anthopogonoides Maxim, a kind of traditional Tibetan medicine, has been used to remove body heat, body detoxification, cough, asthma, stomachic and swelling, eliminate abundant phlegm and inflammatory for a long time. In the present study, the total phenols and total flavonoid contents as well as antioxidative properties of the crude extract and solvent fractions of R. anthopogonoides were determined using seven antioxidant assays. Additionally, the protective effect of the extracts on hypoxia-induced injury in PC12 cells was also investigated. The content of total flavonoid and total phenolic was determined by the aluminum colorimetric method and Folin-Ciocalteu assay, respectively. In vitro antioxidant study, the effect of the crude extract and solvent fractions on total antioxidant activity, reducing power, DPPH radical scavenging, ABTS radical scavenging, superoxide radical scavenging, hydroxyl radical scavenging and nitric oxide radical scavenging were examined. The correlation between the phenolic and flavonoid content of the extracts and their antioxidant properties also analyzed. Furthermore, the protective effect of extracts on hypoxia-induced damage on PC12 cells was investigated by cell viability, lactate dehydrogenase (LDH) release, malondialdehyde (MDA) production and the activities of antioxidant enzymes. Our results showed that ethyl acetate and n-butanol fractions had higher content of phenolics and flavonoid compounds than other fractions. Except ABTS radical assay, n-butanol fraction exhibited the strongest antioxidant activity. While the hexane fraction showed the lowest antioxidant activity. Ethyl acetate also presented excellent antioxidant activity, which was just lower than n-butanol fraction. Significant correlation between the phenolic, flavonoid content of the extract and fractions with antioxidant assay excluding ABTS, OH scavenging assay was observed. Moreover, ethyl acetate and n-butanol fractions showed protective effect in PC12 cell under hypoxia condition, while crude extract and water fraction had no protective effect. In contrast, hexane fraction exhibited strong cytoprotective effect. Further study indicated that pretreatment of PC12 cells with ethyl acetate and n-butanol fractions, prior to hypoxia exposure, significantly increased the survival of cells and the activities of SOD, CAT, GSH-Px and T-AOC, as well as reduced the level of LDH and MDA. The gathered data demonstrated that ethyl acetate and n-butanol fractions were able to protect PC12 cells against hypoxia induced injury through direct free radical scavenging and modulation of endogenous antioxidant enzymes. These findings suggested that ethyl acetate and n-butanol fractions of R. anthopogonoides had significant antioxidant activity and could prevent PC12 cells against hypoxia-induced injury. So it might be regarded as an excellent source of antioxidants and had great potential to explore as therapeutic agent for preventing hypoxia related sickness in future.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
281689
1,1-Diphenyl-2-picrylhydrazine, 97%
C18H13N5O6
D9132
2,2-Diphenyl-1-picrylhydrazyl
C18H12N5O6
T5500
2-Thiobarbituric acid, ≥98%
C4H4N2O2S
32950
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
C13H14N2
277908
Ammonium molybdate, 99.98% trace metals basis
H8MoN2O4
A1343
Ammonium molybdate, meets USP testing specifications
H24Mo7N6O24 · 4H2O
M1019
Ammonium molybdate tetrahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, 81.0-83.0% MoO3 basis
H24Mo7N6O24 · 4H2O
09878
Ammonium molybdate tetrahydrate, BioUltra, ≥99.0% (T)
H24Mo7N6O24 · 4H2O
270989
Ethyl acetate, anhydrous, 99.8%
C4H8O2
437549
Ethyl acetate, ACS reagent, ≥99.5%
C4H8O2
270520
Ethyl acetate, HPLC grade, 99.8%
C4H8O2
W241407
Ethyl acetate, ≥99%, FCC, FG
C4H8O2
09-0703
Ethyl acetate, for residue analysis, JIS 5000
C4H8O2
09-0730
Ethyl acetate, for HPLC
C4H8O2
W241415
Ethyl acetate, natural, ≥99%, FCC, FG
C4H8O2
537446
Ethyl acetate, ReagentPlus®, ≥99.8%
C4H8O2
L092004
Ethyl acetate
C4H8O2
L092003
Ethyl acetate
C4H8O2
G4251
L-Glutathione reduced, ≥98.0%
C10H17N3O6S
G6529
L-Glutathione reduced, BioXtra, ≥98.0%
C10H17N3O6S
G6013
L-Glutathione reduced, cell culture tested, BioReagent, ≥98.0%, powder
C10H17N3O6S
84010
NBT Vials, for in vitro diagnostic use
C40H30Cl2N10O6
N6876
Nitrotetrazolium Blue chloride, ~98% (TLC)
C40H30Cl2N10O6
N6639
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
C40H30Cl2N10O6
P13207
Phenazine, 98%
C12H8N2
P9625
Phenazine methosulfate, ≥90% (UV)
C14H14N2O4S
379824
Potassium persulfate, 99.99% trace metals basis
K2O8S2
190780
Sodium trichloroacetate, 97%
C2Cl3NaO2
S9251
Sulfanilamide, ≥99%
C6H8N2O2S
33626
Sulfanilamide, puriss. p.a., ≥99% (calc. to the dried substance)
C6H8N2O2S