EMAIL THIS PAGE TO A FRIEND
Molecular and Cellular Biology
A Synthetic Lethal Interaction between Glutathione Synthesis and Mitochondrial Reactive Oxygen Species Provides a Tumor-Specific Vulnerability Dependent on STAT3.
PMID 26283727